Learn More
Cayman Chemical 7Z,10Z,13Z,16Z-Docosatetraenoic Acid, 28874-58-0, 25 mg
SDP
Supplier:  Cayman Chemical 9030025
Encompass_Preferred
Molecular Formula C22H36O2, Purity ≥98%, Formula Weight 332.5, Synonyms: 7Z,10Z,13Z,16Z-docosatetraenoic acid, Adrenic Acid; A naturally occurring PUFA formed through a 2-carbon chain elongation of arachidonic acid; present in the adrenal glands, brain, testis, and kidney.
Catalog No. NC1115827