Learn More
Cayman Chemical Adrenic Acid, Purity: ≥98%, CAS: 28874-58-0, 50 mg
SDP
Catalog No. 502895439
Encompass_Preferred
A naturally occurring PUFA formed through a 2-carbon chain elongation of arachidonic acid; present in the adrenal glands, brain, testis, and kidney
Catalog No. 50-289-5439 Supplier Cayman Chemical Supplier No. 9030050 MG
A naturally occurring PUFA formed through a 2-carbon chain elongation of arachidonic acid; present in the adrenal glands, brain, testis, and kidney