Learn More
Medchemexpress LLC Adrenic acid (cis-7,10,13,16-docosatetraenoic acid) | 28874-58-0 | 99.6% | 332.52 | 5 MG
SDP
Catalog No. 5000454445
Encompass_Preferred
Adrenic Acid (cis-7,10,13,16-Docosatetraenoic acid) is a naturally polyunsaturated fatty acid found in the adrenal gland, brain, kidney, and vasculature. It can regulate vascular tone in arteries of the adrenal cortex and acts as an inflammation enhancer in non-alcoholic fatty liver disease. It is produced via the elongation of arachidonic acid by ELOVL2 and 5. This product is for research use only.
  • Naturally polyunsaturated fatty acid
  • Found in adrenal gland, brain, kidney, and vasculature
  • Regulates vascular tone in arteries of the adrenal cortex
  • Acts as an inflammation enhancer in non-alcoholic fatty liver disease
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • For research use only
Catalog No. 50-004-54445 Supplier Medchemexpress LLC Supplier No. HYW0132155MG
Add to Cart
Add to Cart
Adrenic Acid (cis-7,10,13,16-Docosatetraenoic acid) is a naturally polyunsaturated fatty acid found in the adrenal gland, brain, kidney, and vasculature. It can regulate vascular tone in arteries of the adrenal cortex and acts as an inflammation enhancer in non-alcoholic fatty liver disease. It is produced via the elongation of arachidonic acid by ELOVL2 and 5. This product is for research use only.
  • Naturally polyunsaturated fatty acid
  • Found in adrenal gland, brain, kidney, and vasculature
  • Regulates vascular tone in arteries of the adrenal cortex
  • Acts as an inflammation enhancer in non-alcoholic fatty liver disease
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • For research use only