Learn More
Medchemexpress LLC HY-P1518 5mg Medchemexpress, Adrenocorticotropic Hormone (ACTH) (1-10), human CAS:2791-05-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP15185MG
Encompass_Preferred
Medchemexpress, HY-P1518 5mg Adrenocorticotropic Hormone (ACTH) (1-10), human CAS:2791-05-1 Adrenocorticotropic Hormone (ACTH) (1-10), human, an adrenocorticotropin hormone fragment, possesses a weak α-melanocyte stimulating hormone (α-MSH) potency only at high doses (100 and 1000 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4219