Learn More
Sigma Aldrich Fine Chemicals Biosciences Adrenocorticotropic Hormone Fragment 18-39 human | 53917-42-3 | MFCD00076159 | 1mg
Catalog No. NC1811640
Encompass
Adrenocorticotropic Hormone Fragment 18-39 human | Purity: ≥97% (HPLC) | MW: 2465.67 | 53917-42-3 | MFCD00076159 | 1mg
Catalog No. NC1811640 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A06731MG
Adrenocorticotropic Hormone Fragment 18-39 human | Purity: ≥97% (HPLC) | MW: 2465.67 | 53917-42-3 | MFCD00076159 | 1mg