Learn More
Carbosynth LLC. Adrenomedullin 2 (Human) (0.1mg vial), 1ea, Disulfide bonds: (Disulfide bond between Cys10 and Cys15), MW: 5100.7, Molecular formula: C219H349N69O66S3
SDP
Supplier:  Carbosynth LLC. PAD4421S0.1MG
Encompass_Preferred
Synonyms: Thr-Gln-Ala-Gln-Leu-Leu-Arg-Val-Gly-Cys-Val-Leu-Gly-Thr-Cys-Gln-Val-Gln-Asn-Leu-Ser-His-Arg-Leu-Trp-Gln-Leu-Met-Gly-Pro-Ala-Gly-Arg-Gln-Asp-Ser-Ala-Pro-Val-Asp-Pro-Ser-Ser-Pro-His-Ser-Tyr-NH2, Intermedin (Human), H-TQAQLLRVGCVLGTCQVQNLSHRLWQLMGPAGRQDSAPVDPSSPHSY-NH2, Application: Potent Cardiovascular and Renal Regulator / Suppressor for Food Intake and Gastric Emptying, Storage Temperature: -20°C, References: M. Ogoshi, K. Inoue, and Y. Takei, Biochem. Biophys. Res. Commun., 311, 1072 (2003).(Takifugu rubripes Adrenomedullins), Y. Takei, K. Inoue, M. Ogoshi, T. Kawahara, H. Bannai, and S. Miyano, FEBS Lett., 556, 53 (2004). (Original; Adrenomedullin 2), J. Roh, C.L. Chang, A. Bhalla, C. Klein, and S.Y.T. Hsu, J. Biol. Chem., 279, 7264 (2004). (Original; Intermedin)
Catalog No. 50-168-6103