Learn More
Bachem Adrenomedullin (22-52) (human) Trifluoroacetate
SDP
Supplier:  Bachem 4028071.0500
Encompass_Preferred
Adrenomedullin (22-52) (human) Trifluoroacetate, H-Thr-Val-Gln-Lys-Leu-Ala-His-Gln-Ile-Tyr-Gln-Phe-Thr-Asp-Lys-Asp-Lys-Asp-Asn-Val-Ala-Pro-Arg-Ser-Lys-Ile-Ser-Pro-Gln-Gly-Tyr-NH2, 159899-65-7, C159H252N46O48, Mr 3576.03, 0.5mg. ADM (22-52) is a putative adrenomedullin receptor antagonist that inhibits the production of cAMP elicted by adrenomedullin. It is a more effective antagonist for adrenomedullin- and CGRP-specific receptors than a-CGRP (8-37) (human).
Catalog No. 50-382-896