Learn More
Bachem Adrenomedullin (human) Trifluoroacetate
SDP
Supplier:  Bachem 4030315.1000
Encompass_Preferred
Adrenomedullin (human) Trifluoroacetate, H-Tyr-Arg-Gln-Ser-Met-Asn-Asn-Phe-Gln-Gly-Leu-Arg-Ser-Phe-Gly-Cys-Arg-Phe-Gly-Thr-Cys-Thr-Val-Gln-Lys-Leu-Ala-His-Gln-Ile-Tyr-Gln-Phe-Thr-Asp-Lys-Asp-Lys-Asp-Asn-Val-Ala-Pro-Arg-Ser-Lys-Ile-Ser-Pro-Gln-Gly-Tyr-NH2, 148498-78-6, C264H406N80O77S3, Mr 6028.82, 1mg. ADM (1-52) (human), Adrenomedullin (1-52), human. Adrenomedullin (ADM) elicits a potent and longlasting hypotensive effect and is considered as a hormone involved in blood pressure control. Additionally, ADM acts as a protective factor of blood vessels. Adrenomedullin is a potent stimulator of osteoblastic activity. Further biological activities of hADM include reduction of oxidative stress and inhibition of endothelial cell apoptosis.
Catalog No. NC9907472