Learn More
CPC Scientific H-Thr-Val-Gln-Lys-Leu-Ala-His-Gln-Ile-Tyr-Gln-Phe-Thr-Asp-Lys-Asp-Lys-Asp-Asn-Val-Ala-Pro-Arg-Ser-Lys-Ile-Ser-Pro-Gln-Gly-Tyr-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ADRM005B
Encompass
Adrenomedullin-elicted cAMP antagonist and a more effective inhibitor than alpha-CGRP in terms of adrenomedullin- and CGRP-specific receptors.ONE-LETTER SEQUENCE: TVQKLAHQIYQFTDKDKDNVAPRSKISPQGY-NH2MOLECULAR FORMULA: C159H252N46O48MOLECULAR WEIGHT:3576.1STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [159899-65-7], RESEARCH AREA: Cardiovascular
Catalog No. 50-210-8624