Learn More
Apexbio Technology LLC Adrenorphin, Free Acid 88866-92-6 10mg
SDP
Catalog No. 5000095217
Encompass_Preferred
Adrenorphin Free Acid is an endogenous opioid peptide agonist targeting mu ( ) and kappa ( ) opioid receptors It is designed to modulate opioid receptor signaling thereby regulating analgesic and respiratory modulatory pathways in mammalian systems Adrenorphin Free Acid exerts its biological activity primarily through selective activation of and opioid receptors without activity at delta ( ) receptors Based on these pharmacological properties Adrenorphin Free Acid holds research potential in investigating opioid receptor signaling mechanisms pain modulation processes and opioid-related respiratory effects in biomedical research
Catalog No. 50-000-95217 Supplier Apexbio Technology LLC Supplier No. A101710
Add to Cart
Add to Cart
Adrenorphin Free Acid is an endogenous opioid peptide agonist targeting mu ( ) and kappa ( ) opioid receptors It is designed to modulate opioid receptor signaling thereby regulating analgesic and respiratory modulatory pathways in mammalian systems Adrenorphin Free Acid exerts its biological activity primarily through selective activation of and opioid receptors without activity at delta ( ) receptors Based on these pharmacological properties Adrenorphin Free Acid holds research potential in investigating opioid receptor signaling mechanisms pain modulation processes and opioid-related respiratory effects in biomedical research