Learn More
Apexbio Technology LLC Adrenosterone 382-45-6 1g
SDP
Catalog No. 5000092547
Encompass_Preferred
Adrenosterone (CAS 382-45-6) is a small-molecule inhibitor targeting 11- -hydroxysteroid dehydrogenase 1 It is designed to competitively inhibit this enzyme thereby modulating steroid metabolism pathways Adrenosterone exerts its biological activity primarily through enzymatic inhibition In human cancer cell studies Adrenosterone demonstrates inhibitory activity on metastatic progression Based on these pharmacological properties Adrenosterone holds research potential in studying steroid metabolism androgen receptor interactions endocrine regulation mechanisms and hormone-dependent diseases
Catalog No. 50-000-92547 Supplier Apexbio Technology LLC Supplier No. B10421000
Adrenosterone (CAS 382-45-6) is a small-molecule inhibitor targeting 11- -hydroxysteroid dehydrogenase 1 It is designed to competitively inhibit this enzyme thereby modulating steroid metabolism pathways Adrenosterone exerts its biological activity primarily through enzymatic inhibition In human cancer cell studies Adrenosterone demonstrates inhibitory activity on metastatic progression Based on these pharmacological properties Adrenosterone holds research potential in studying steroid metabolism androgen receptor interactions endocrine regulation mechanisms and hormone-dependent diseases