Learn More
Medchemexpress LLC HY-19705B 5mg Medchemexpress, Adriforant (hydrochloride) CAS:2096455-90-0 Purity:>98%
SDP
Catalog No. 502082165
Encompass_Preferred
Medchemexpress, HY-19705B 5mg Adriforant (hydrochloride) CAS:2096455-90-0 Adriforant hydrochloride (PF-3893787 hydrochloride) is a novel histamine H4 receptor antagonist binding affinity (Ki=2.4 nM) and is also a functional (Ki=1.56 nM) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2165 Supplier Medchemexpress LLC Supplier No. HY19705B5MG
Medchemexpress, HY-19705B 5mg Adriforant (hydrochloride) CAS:2096455-90-0 Adriforant hydrochloride (PF-3893787 hydrochloride) is a novel histamine H4 receptor antagonist binding affinity (Ki=2.4 nM) and is also a functional (Ki=1.56 nM) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.