Learn More
Medchemexpress LLC Adriforant hydrochloride (PF-3893787 hydrochloride) | 2096455-90-0 | 98.0% | 371.74 | 1 ML
SDP
Catalog No. 5000401327
Encompass_Preferred
Adriforant hydrochloride (PF-3893787 hydrochloride) is a novel histamine H4 receptor antagonist with a binding affinity (Ki) of 2.4 nM and is also a functional (Ki) antagonist at 1.56 nM. It is suitable for laboratory research and studies involving histamine receptors.
  • Novel histamine H4 receptor antagonist
  • High binding affinity and functional antagonism
  • Suitable for high-speed applications
  • Leak-resistant cap for secure storage
Catalog No. 50-004-01327 Supplier Medchemexpress LLC Supplier No. HY19705B10MM/1ML
Adriforant hydrochloride (PF-3893787 hydrochloride) is a novel histamine H4 receptor antagonist with a binding affinity (Ki) of 2.4 nM and is also a functional (Ki) antagonist at 1.56 nM. It is suitable for laboratory research and studies involving histamine receptors.
  • Novel histamine H4 receptor antagonist
  • High binding affinity and functional antagonism
  • Suitable for high-speed applications
  • Leak-resistant cap for secure storage