Learn More
Medchemexpress LLC Adriforant hydrochloride (PF-3893787 hydrochloride) | 2096455-90-0 | 98.0% | 371.74 | 100 MG
SDP
Catalog No. 5000400885
Encompass_Preferred
Adriforant hydrochloride (PF-3893787 hydrochloride) is a novel histamine H4 receptor antagonist. It is for research use only and not sold to patients. This product is suitable for laboratory research in various biological assays.
  • Novel histamine H4 receptor antagonist
  • High binding affinity (Ki=2.4 nM)
  • Functional antagonist (Ki=1.56 nM)
  • Tested and observed binding Ki = 2.4 nM and functional Ki = 1.56 nM for H4R
  • In vitro IC50 on human native isolated eosinophils assessing actin polymerisation is 1.16 nM
  • Used in clinical trials for psoriasis and atopic dermatitis
Catalog No. 50-004-00885 Supplier Medchemexpress LLC Supplier No. HY19705B100MG
Adriforant hydrochloride (PF-3893787 hydrochloride) is a novel histamine H4 receptor antagonist. It is for research use only and not sold to patients. This product is suitable for laboratory research in various biological assays.
  • Novel histamine H4 receptor antagonist
  • High binding affinity (Ki=2.4 nM)
  • Functional antagonist (Ki=1.56 nM)
  • Tested and observed binding Ki = 2.4 nM and functional Ki = 1.56 nM for H4R
  • In vitro IC50 on human native isolated eosinophils assessing actin polymerisation is 1.16 nM
  • Used in clinical trials for psoriasis and atopic dermatitis