Learn More
Aviva Systems Biology Corp ADRM1 Antibody - C-terminal region (ARP59363_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59363_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADRM1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ADRM1. Alternate names: ARM1, GP110, MGC29536, Rpn13ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rat
Catalog No. 50-286-674