Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci ADRM1 Peptide
SDP

Catalog No. 16595965
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-965 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-965 Supplier ProSci Supplier No. 8119P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

ADRM1 peptide is used for blocking the activity of ADRM1 antibody.

Specifications

Gene ID (Entrez) 11047
Species Human
Accession Number NP_783163
Common Name adhesion regulating molecule 1
Concentration 200 μg/mL
Content And Storage Store ADRM1 peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol ADRM1
Quantity 0.05 mg
Sequence 19 amino acid peptide near the center of human ADRM1.
Synonym Adhesion regulating molecule 1, ARM1, Gp110, hRpn13, Proteasomal ubiquitin receptor ADRM1, Proteasome regulatory particle non-ATPase 13, Rpn13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.