Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adrogolide hydrochloride | 166591-11-3 | 99.6% | 435.96 | C22H26ClNO4S | 100 MG
SDP

Catalog No. 5000307529
Encompass_Preferred

Adrogolide hydrochloride is a chemically stable prodrug that is converted rapidly in plasma to the dopamine D1 receptor agonist A-86929 and is used in preclinical research on Parkinson's disease and dopaminergic signaling. It is supplied as a white to off-white solid with documented purity, molecular weight, solubility, and storage recommendations.

  • High purity: 99.6% (reported)
  • Molecular weight 435.96
  • Chemical formula C22H26ClNO4S
  • Soluble in DMSO (≈66.7 mg/mL with warming)
  • Documented in vivo formulations for vehicle preparation
  • Stable prodrug that converts to A-86929 in plasma
  • Store at room temperature; in solvent store at -80°C for long term

Catalog No. 50-003-07529 Supplier Medchemexpress LLC Supplier No. HY118956100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Adrogolide hydrochloride is a chemically stable prodrug that is converted rapidly in plasma to the dopamine D1 receptor agonist A-86929 and is used in preclinical research on Parkinson's disease and dopaminergic signaling. It is supplied as a white to off-white solid with documented purity, molecular weight, solubility, and storage recommendations.

  • High purity: 99.6% (reported)
  • Molecular weight 435.96
  • Chemical formula C22H26ClNO4S
  • Soluble in DMSO (≈66.7 mg/mL with warming)
  • Documented in vivo formulations for vehicle preparation
  • Stable prodrug that converts to A-86929 in plasma
  • Store at room temperature; in solvent store at -80°C for long term

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.