Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adrogolide hydrochloride | 166591-11-3 | 99.6% | 435.96 | C22H26ClNO4S | 25 MG
SDP

Catalog No. 5000227534
Encompass_Preferred

Adrogolide hydrochloride is a research-grade prodrug converted rapidly in plasma to the dopamine D1 receptor agonist A-86929. Supplied as the hydrochloride salt, it is intended for laboratory research use only and is provided in pre-weighed quantities for assay preparation.

  • High purity (≈99.6%) suitable for research applications.
  • Molecular weight 435.96 and formula C22H26ClNO4S.
  • Available in pre-weighed small quantities for in vitro and in vivo studies.
  • Soluble in DMSO for assay preparation.
  • Acts as a prodrug to the D1 receptor agonist A-86929.

Catalog No. 50-002-27534 Supplier Medchemexpress LLC Supplier No. HY11895625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adrogolide hydrochloride is a research-grade prodrug converted rapidly in plasma to the dopamine D1 receptor agonist A-86929. Supplied as the hydrochloride salt, it is intended for laboratory research use only and is provided in pre-weighed quantities for assay preparation.

  • High purity (≈99.6%) suitable for research applications.
  • Molecular weight 435.96 and formula C22H26ClNO4S.
  • Available in pre-weighed small quantities for in vitro and in vivo studies.
  • Soluble in DMSO for assay preparation.
  • Acts as a prodrug to the D1 receptor agonist A-86929.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.