Learn More
Medchemexpress LLC Adrogolide hydrochloride | 166591-11-3 | 99.6% | 435.96 g·mol⁻¹ | C22H26ClNO4S | 10 MG
SDP
Catalog No. 5000307530
Encompass_Preferred
Adrogolide hydrochloride is a research prodrug that is converted rapidly in plasma to A-86929, a dopamine D1 receptor agonist used in preclinical neuropharmacology studies. It has been studied in Parkinson's disease models for effects on motor function and dyskinesia.
  • Research prodrug to a dopamine D1 receptor agonist for pharmacology studies.
  • Used in preclinical Parkinson's disease and dyskinesia models.
  • High analytical purity (99.6%).
  • Available in small-mass research pack sizes suitable for in vitro and in vivo studies.
  • Soluble in DMSO; forms clear solutions at ≥ 2.5 mg/mL.
Catalog No. 50-003-07530 Supplier Medchemexpress LLC Supplier No. HY11895610MG
Adrogolide hydrochloride is a research prodrug that is converted rapidly in plasma to A-86929, a dopamine D1 receptor agonist used in preclinical neuropharmacology studies. It has been studied in Parkinson's disease models for effects on motor function and dyskinesia.
  • Research prodrug to a dopamine D1 receptor agonist for pharmacology studies.
  • Used in preclinical Parkinson's disease and dyskinesia models.
  • High analytical purity (99.6%).
  • Available in small-mass research pack sizes suitable for in vitro and in vivo studies.
  • Soluble in DMSO; forms clear solutions at ≥ 2.5 mg/mL.