Learn More
Medchemexpress LLC Adrogolide hydrochloride | 166591-11-3 | 99.6% | 435.96 g/mol | C22H26ClNO4S | 5 MG
SDP
Catalog No. 5000307528
Encompass_Preferred
Adrogolide hydrochloride is the hydrochloride salt of adrogolide, a prodrug that is converted in plasma to the dopamine D1 receptor agonist A-86929. It is provided as a research-grade compound for pharmacology and neuroscience studies where selective D1 activation is required.
  • High purity suitable for research applications.
  • Soluble in DMSO; documented in vivo formulation options for administration.
  • Available in small and larger pack sizes to support screening and in vivo studies.
  • Stable when stored dry at room temperature; stock solutions stable at -20°C to -80°C.
  • Used to study dopamine D1 receptor-mediated pharmacology in vitro and in vivo.
Catalog No. 50-003-07528 Supplier Medchemexpress LLC Supplier No. HY1189565MG
Adrogolide hydrochloride is the hydrochloride salt of adrogolide, a prodrug that is converted in plasma to the dopamine D1 receptor agonist A-86929. It is provided as a research-grade compound for pharmacology and neuroscience studies where selective D1 activation is required.
  • High purity suitable for research applications.
  • Soluble in DMSO; documented in vivo formulation options for administration.
  • Available in small and larger pack sizes to support screening and in vivo studies.
  • Stable when stored dry at room temperature; stock solutions stable at -20°C to -80°C.
  • Used to study dopamine D1 receptor-mediated pharmacology in vitro and in vivo.