Learn More
Medchemexpress LLC Adropin (34-76) (human, mouse, rat) | 1802086-30-1 | 98.2% | 4499.82 | 1 MG
SDP
Catalog No. 5000398420
Encompass_Preferred
Adropin (34-76) is a secretory domain of Adropin. It can inhibit cAMP levels and glucose production in hepatocytes, exhibiting a hypoglycemic effect. This peptide also plays an antifibrotic role by inhibiting the GLI1 signaling pathway.
  • Inhibits glucose production in primary cultured mouse hepatocytes.
  • Decreases cAMP levels in HepG2 hepatocytes.
  • Inhibits TGF-β-induced cell activation, collagen production, and fibrotic remodeling in fibroblasts.
  • Plays an anti-fibrotic role by inhibiting the GLI1 signaling pathway.
  • Has a hypoglycemic effect in a diet-induced obesity model of mice.
  • Shows improved effect in mouse models of fibrosis.
Catalog No. 50-003-98420 Supplier Medchemexpress LLC Supplier No. HYP48601MG
Adropin (34-76) is a secretory domain of Adropin. It can inhibit cAMP levels and glucose production in hepatocytes, exhibiting a hypoglycemic effect. This peptide also plays an antifibrotic role by inhibiting the GLI1 signaling pathway.
  • Inhibits glucose production in primary cultured mouse hepatocytes.
  • Decreases cAMP levels in HepG2 hepatocytes.
  • Inhibits TGF-β-induced cell activation, collagen production, and fibrotic remodeling in fibroblasts.
  • Plays an anti-fibrotic role by inhibiting the GLI1 signaling pathway.
  • Has a hypoglycemic effect in a diet-induced obesity model of mice.
  • Shows improved effect in mouse models of fibrosis.