Learn More
TARGETMOL CHEMICALS INC Adropin 34-76 human mouse rat
SDP
Catalog No. 502876583
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Adropin (34-76) (human, mouse, rat) is a peptide that attenuates liver fibrosis and insulin resistance independent of obesity or food intake. Purity 99.95%
Catalog No. 50-287-6583 Supplier TARGETMOL CHEMICALS INC Supplier No. T766641MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Adropin (34-76) (human, mouse, rat) is a peptide that attenuates liver fibrosis and insulin resistance independent of obesity or food intake. Purity 99.95%