Learn More
Surgical Design™ Adson Aneurysm Needle
Catalog No. 21124622 Shop All Surgical Design Products
Change view
Click to view available options
Length (English):
6.5 in.
Catalog No. Length (English)
21-124-622 6.5 in.
1 options
Catalog No. 21-124-622 Supplier Surgical Design™ Supplier No. ATPM002
Add to Cart
Add to Cart
Adson aneurysm needle, 9 in., features a blunt angled tip with eyelet for vessel isolation, tissue elevation, and controlled postmortem dissection.
  • Manufactured from premium surgical-grade stainless steel, durable for repeated pathology and mortuary lab use
  • Extra-long 9-in. design provides extended reach for deep thoracic, abdominal, and cranial cavity exploration
  • Blunt, curved aneurysm tip prevents accidental perforation of vessels or soft tissues during delicate postmortem dissection
  • Integrated eyelet ideal for passing twine, ligatures, or vessel loops to isolate and secure arteries during organ removal
  • Slim, tapered shaft enhances visibility and maneuverability when exposing anatomical pathways
  • Textured grip ensures stability when working in moist autopsy environments
  • Satin-finished surface reduces glare under strong exam lighting
  • Autoclavable and fully reusable, engineered for forensic, pathology, anatomy, and mortuary settings
  • Isolating vessels prior to organ extraction
  • Guiding ligatures around arteries and veins
  • Lifting tissues to assist with cardiovascular and neck dissections
  • Mapping vascular routes in forensic investigations
  • Suitable for full, partial, and forensic autopsies as well as anatomy labs
Specifications
Product Type Needle Probe
Autoclavable Autoclavable
Certifications/Compliance ISO/GMP
Color Silver
Material Stainless Steel
Latex Free Yes
Length (English) 6.5 in.
Sterility Non-sterile