Learn More
Electron Microscopy Sciences Adson Bone Rongeur, Slightly Curved
SDP
Catalog No. 5030184
Encompass_Preferred
Made from stainless steel with a polished finish. Slightly curved jaws.
OAL: 8"
Jaw width: 7mm
Catalog No. 50-301-84 Supplier Electron Microscopy Sciences Supplier No. 77985C
Made from stainless steel with a polished finish. Slightly curved jaws.
OAL: 8"
Jaw width: 7mm