Learn More
Sklar Instruments Sklarlite XD Adson Dressing Forceps, Sklar, 4-3/4 in., Straight, Serrated, Extra Light and Extra Delicate (SklarLite XD), Stainless Steel. EA/1.
SDP
Catalog No. 501437987 Shop All Sklar Instruments Products
Encompass_Preferred
Sklarlite XD Adson Dressing Forceps, Sklar, 4-3/4 in., Straight, Serrated, Extra Light and Extra Delicate (SklarLite XD), Stainless Steel. Sklarlite XD Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They have a wide thumb grasp for increased precision and control. Sklarlite XD instruments are noticeably lighter and delicate than standard instruments which allow the surgeon the highest degree of precision with the least amount of fatigue. These instruments have been uniquely manufactured to be one of the lightest, most delicate general instruments available today. This product is straight with serrated tips and a length of 4-3/4 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7987 Supplier Sklar Instruments Supplier No. 232604
Sklarlite XD Adson Dressing Forceps, Sklar, 4-3/4 in., Straight, Serrated, Extra Light and Extra Delicate (SklarLite XD), Stainless Steel. Sklarlite XD Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They have a wide thumb grasp for increased precision and control. Sklarlite XD instruments are noticeably lighter and delicate than standard instruments which allow the surgeon the highest degree of precision with the least amount of fatigue. These instruments have been uniquely manufactured to be one of the lightest, most delicate general instruments available today. This product is straight with serrated tips and a length of 4-3/4 inches.