Learn More
Sklar Instruments Forceps, Merit Adson-Brown Tissue Forceps, Sklar 4-3/4 in., 7x7 Teeth Side-Grasping, Straight, Thumb, Stainless Steel, EA.
SDP
Catalog No. 501126687 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Merit Adson-Brown Tissue Forceps, Sklar, 4-3/4 in., 7x7 Teeth Side-Grasping, Straight, Thumb, Stainless Steel, EA. Merit Adson-Brown Tissue forceps are for holding and manipulating delicate tissues. These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow. They are similar to standard Adson forceps, however, Adson-Brown forceps feature multiple rows of teeth, typically 7x7 or 9x9. They are used to grasp delicate tissue, most commonly in plastic surgery. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit instruments deliver an economical solution for physicians and practices that require quality, durable instruments.
Catalog No. 50-112-6687 Supplier Sklar Instruments Supplier No. 97778
Forceps, Merit Adson-Brown Tissue Forceps, Sklar, 4-3/4 in., 7x7 Teeth Side-Grasping, Straight, Thumb, Stainless Steel, EA. Merit Adson-Brown Tissue forceps are for holding and manipulating delicate tissues. These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow. They are similar to standard Adson forceps, however, Adson-Brown forceps feature multiple rows of teeth, typically 7x7 or 9x9. They are used to grasp delicate tissue, most commonly in plastic surgery. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit instruments deliver an economical solution for physicians and practices that require quality, durable instruments.