Learn More
Fine Science Tools ADSON DBAKEY FORCEPS 15CM
SDP
Catalog No. NC1855937
Encompass
Tip Shape:Straight; Tips:DeBakey Serrations; Tip Width:1.5mm; Tip Dimensions:1.75 x 1.5mm; Alloy / Material:Stainless Steel;Length:15cm; FINE SCIENCE TOOLS INC mfg# 11616-15
Catalog No. NC1855937 Supplier Fine Science Tools Supplier No. 1161615
Tip Shape:Straight; Tips:DeBakey Serrations; Tip Width:1.5mm; Tip Dimensions:1.75 x 1.5mm; Alloy / Material:Stainless Steel;Length:15cm; FINE SCIENCE TOOLS INC mfg# 11616-15