Learn More
Sklar Instruments Econo Adson Dressing Forceps, Sklar, 4-3/4 in., Blunt, Straight, Serrated, Thumb, Stainless Steel. PK/12.
SDP
Catalog No. 501437681 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Adson Dressing Forceps, Sklar, 4-3/4 in., Blunt, Straight, Serrated, Thumb, Stainless Steel. Econo Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They have a wide thumb grasp for increased precision and control. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with serrated tips and a length of 4-3/4 inches. They are sold in boxes of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7681 Supplier Sklar Instruments Supplier No. 21771
Econo Adson Dressing Forceps, Sklar, 4-3/4 in., Blunt, Straight, Serrated, Thumb, Stainless Steel. Econo Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They have a wide thumb grasp for increased precision and control. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with serrated tips and a length of 4-3/4 inches. They are sold in boxes of 12.