Learn More
Surgical Design™ Adson Dressing Forceps, Serrated, Fenestrated Handle
Catalog No. 21124574 Shop All Surgical Design Products
Change view
Click to view available options
Color:
Black
Blue
Multi-colored
Silver
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-124-574 Multi-colored
21-124-571 Silver
21-124-572 Black
21-124-573 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-124-574 Supplier Surgical Design™ Supplier No. 249NPR
Add to Cart
Add to Cart
Adson dressing forceps with a fenestrated handle provide precision for secure dressing placement and delicate tissue handling.
  • Constructed from premium surgical-grade stainless steel for precision, balance, and long-lasting durability
  • Fine serrated tips ensure secure grip on dressings, gauze, and delicate tissue without slippage or damage
  • Fenestrated handle design reduces instrument weight while providing improved control and comfort
  • Available in four premium polished finishes — Silver, Black, Blue, and Multicolor Titanium
  • Non-slip handle texture enhances grip even during wet or gloved operation
  • Corrosion-resistant finish for long-lasting shine and easy sterilization
  • Lightweight, ergonomic construction minimizes hand fatigue in prolonged use
  • Autoclavable and reusable, built for surgical, clinical, and dental environments
  • Precision alignment for smooth, reliable performance over repeated sterilization cycles
  • Ideal for dressing application and removal, tissue manipulation, and suture assistance
  • Commonly used in general surgery, plastic surgery, veterinary, and clinical procedures
  • Excellent for medical kits, teaching labs, and first aid use
Specifications
Product Type Thumb Forceps
Autoclavable Autoclavable
Certifications/Compliance ISO, GMP
Color Multi-colored
Material Stainless Steel
Disposable Reusable
Latex Free Yes
Style Dressing
Teeth Count Serrated
Length (English) 4.75 in.
Length (Metric) 12 cm
Sterility Non-sterile