Learn More
Sklar Instruments Adson (Ewald) Dressing Forceps, 4-3/4 in., Straight, Serrated, Stainless Steel, EA.
SDP
Catalog No. 501109333 Shop All Sklar Instruments Products
Encompass_Preferred
Adson (Ewald) Dressing Forceps, 4-3/4 in., Straight, Serrated, Stainless Steel, EA. Sklar's Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They have a wide thumb grasp for increased precision and control. This product may also be known as Ewald forceps. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. This product is straight with serrated edges, and a length of 4-3/4 inches.
Catalog No. 50-110-9333 Supplier Sklar Instruments Supplier No. 503147
Adson (Ewald) Dressing Forceps, 4-3/4 in., Straight, Serrated, Stainless Steel, EA. Sklar's Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They have a wide thumb grasp for increased precision and control. This product may also be known as Ewald forceps. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. This product is straight with serrated edges, and a length of 4-3/4 inches.