Learn More
Sklar Instruments Adson Dressing Forceps, 4-3/4 in., Straight, Serrated, Delicate, EA.
SDP
Catalog No. 501109332 Shop All Sklar Instruments Products
Encompass_Preferred
Adson Dressing Forceps, 4-3/4 in., Straight, Serrated, Delicate, EA. Sklar Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They have a wide thumb grasp for increased precision and control. This product is straight with delicate, serrated tips and a length of 4-3/4 inches.
Catalog No. 50-110-9332 Supplier Sklar Instruments Supplier No. 472147
Adson Dressing Forceps, 4-3/4 in., Straight, Serrated, Delicate, EA. Sklar Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They have a wide thumb grasp for increased precision and control. This product is straight with delicate, serrated tips and a length of 4-3/4 inches.