Learn More
Roboz Surgical ADSON FCPS 4.75IN 2X3
SDP
Catalog No. NC9446126
Encompass
Adson Forceps; 2X3 Teeth; 1.3mm Tip Width; 4.75 in. Length
Catalog No. NC9446126 Supplier Roboz Surgical Supplier No. RS5234
Adson Forceps; 2X3 Teeth; 1.3mm Tip Width; 4.75 in. Length