Learn More
World Precision Instrument Adson Fine Dressing Forceps, 18 cm, Straight
SDP
Catalog No. 502538314
Encompass
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
Catalog No. 50-253-8314 Supplier World Precision Instrument Supplier No. WP3618
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.