Learn More
World Precision Instrument Adson Forceps, 12cm, 1x2 teeth, 12-pack, Disposable
SDP
Catalog No. 502539447
Encompass
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
Catalog No. 50-253-9447 Supplier World Precision Instrument Supplier No. 50372012
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.