Learn More
World Precision Instrument Adson Forceps, 12cm, Straight, Serrated, 12-pack, Disposable
SDP
Catalog No. 502539448
Encompass
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
Catalog No. 50-253-9448 Supplier World Precision Instrument Supplier No. 50371912
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.