Learn More
World Precision Instrument Adson Forceps, 12cm, Straight, 1mm Serrated Tip, Titanium
SDP
Catalog No. 502538309
Encompass
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.
Catalog No. 50-253-8309 Supplier World Precision Instrument Supplier No. WP4152
Adson forceps have a toothed tip for working with dense tissue, such as skin closures.