Learn More
Fine Science Tools Student Adson Forceps, Stainless Steel, Straight, 12 cm
SDP
Catalog No. NC9017215
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 1.5 x 1mm/ Alloy / Material: Student Stainless Steel/ Length: 12cm/ Feature: Student Quality
Catalog No. NC9017215 Supplier Fine Science Tools Supplier No. 9110612
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 1.5 x 1mm/ Alloy / Material: Student Stainless Steel/ Length: 12cm/ Feature: Student Quality