Learn More
Roboz Surgical ADSON FRCPS 4.75INSERRATED
SDP
Catalog No. NC9495182
Encompass
Adson Forceps; Cross Serrated; Carbide Jaws; 4.75 in. Length; 1mm Tip Width
Catalog No. NC9495182 Supplier Roboz Surgical Supplier No. RS5236
Adson Forceps; Cross Serrated; Carbide Jaws; 4.75 in. Length; 1mm Tip Width