Learn More
Surgical Design™ Adson Tissue Forceps, 1 x 2 Teeth, Fenestrated
Catalog No. 21124565 Shop All Surgical Design Products
Change view
Click to view available options
Color:
Black
Blue
Multi-colored
Silver
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-124-565 Silver
21-124-566 Black
21-124-567 Blue
21-124-568 Multi-colored
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-124-565 Supplier Surgical Design™ Supplier No. 248NP
Only null left
Add to Cart
Add to Cart
Adson tissue forceps with a fenestrated handle deliver grip and control for precision handling.
  • Made from premium surgical-grade stainless steel for exceptional strength, balance, and corrosion resistance
  • 1x2 fine teeth ensure secure tissue grasping with minimal trauma
  • Fenestrated handle design reduces instrument weight while enhancing grip and control
  • Available in four polished finishes — Silver, Black, Blue, and Multicolor Titanium — for style and easy identification
  • Precision-ground tips ensure accurate tissue handling and suture manipulation
  • Non-slip serrations enhance tactile feedback during delicate procedures
  • Ergonomic handle minimizes hand fatigue during long operations
  • Autoclavable and reusable, built for repeated use
  • Mirror-polished resist corrosion and staining
  • Commonly used in plastic surgery, dental, ENT, and microsurgical applications
  • Ideal for tissue manipulation, suture handling, and fine dissection
  • Suitable for operating rooms, laboratories, and teaching facilities
Specifications
Product Type Tissue Forceps
Autoclavable Autoclavable
Certifications/Compliance ISO, GMP
Color Silver
Material Stainless Steel
Disposable Reusable
Latex Free Yes
Style Tissue Forceps
Teeth Count 1 x 2
Length (English) 4.75 in.
Length (Metric) 12 cm
Sterility Non-sterile