Learn More
Sklar Instruments Econo Adson-Brown Tissue Forceps, Sklar, 4-3/4 in., 7x7 Teeth Side-Grasping, Tissue Forceps, Straight, Coarse Teeth, Stainless Steel. PK/12.
SDP
Catalog No. 501437685 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Adson-Brown Tissue Forceps, Sklar, 4-3/4 in., 7x7 Teeth Side-Grasping, Tissue Forceps, Straight, Coarse Teeth, Stainless Steel. Econo Adson-Brown Tissue Forceps are for holding and manipulating delicate tissues. These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow. They are similar to standard Adson forceps, however, Adson-Brown forceps feature multiple rows of teeth, typically 7x7 or 9x9. They are used to grasp delicate tissue, most commonly in plastic surgery. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with 7x7 teeth, and a length of 4-3/4 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7685 Supplier Sklar Instruments Supplier No. 21778
Econo Adson-Brown Tissue Forceps, Sklar, 4-3/4 in., 7x7 Teeth Side-Grasping, Tissue Forceps, Straight, Coarse Teeth, Stainless Steel. Econo Adson-Brown Tissue Forceps are for holding and manipulating delicate tissues. These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow. They are similar to standard Adson forceps, however, Adson-Brown forceps feature multiple rows of teeth, typically 7x7 or 9x9. They are used to grasp delicate tissue, most commonly in plastic surgery. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with 7x7 teeth, and a length of 4-3/4 inches.