Learn More
Sklar Instruments Econo Adson Tissue Forceps, Sklar, 4-3/4 in., 1x2 Teeth, Straight, Smooth, Stainless Steel. PK/12.
SDP
Catalog No. 501437683 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Adson Tissue Forceps, Sklar, 4-3/4 in., 1x2 Teeth, Straight, Smooth, Stainless Steel. Econo Adson Tissue Forceps are for holding and manipulating delicate tissues. These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with smooth jaws, 1 X 2 teeth, and a length of 4-3/4 inches. They are sold in boxes of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7683 Supplier Sklar Instruments Supplier No. 21775
Add to Cart
Add to Cart
Econo Adson Tissue Forceps, Sklar, 4-3/4 in., 1x2 Teeth, Straight, Smooth, Stainless Steel. Econo Adson Tissue Forceps are for holding and manipulating delicate tissues. These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with smooth jaws, 1 X 2 teeth, and a length of 4-3/4 inches. They are sold in boxes of 12.