Learn More
Sklar Instruments Adson Tissue Forceps, 4-3/4 in., 1x2 Teeth, Straight, Smooth, Delicate, Stainless Steel, EA.
SDP
Catalog No. 501109370 Shop All Sklar Instruments Products
Encompass_Preferred
Adson Tissue Forceps, 4-3/4 in., 1x2 Teeth, Straight, Smooth, Delicate, Stainless Steel, EA. Sklar Adson Tissue Forceps are for holding and manipulating delicate tissues. These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow. This product is straight with smooth jaws, delicate 1x2 teeth, and a length of 4-3/4 inches.
Catalog No. 50-110-9370 Supplier Sklar Instruments Supplier No. 503047
Adson Tissue Forceps, 4-3/4 in., 1x2 Teeth, Straight, Smooth, Delicate, Stainless Steel, EA. Sklar Adson Tissue Forceps are for holding and manipulating delicate tissues. These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow. This product is straight with smooth jaws, delicate 1x2 teeth, and a length of 4-3/4 inches.