Learn More
Med Vet International Miltex Vantage Adson Tissue Forceps, 4.75in, with 1x2 Teeth
Supplier:  Med Vet International MILV96120
Encompass_Preferred
The Miltex Vantage V96-120 Adson tissue forceps are made from high quality Japanese stainless steel and measure 4.75' in length. They have a non-locking thumb handle and straight tips that are smooth and feature 1 x 2 teeth.
Catalog No. 50-255-9459
 Restricted Item