Learn More
Fine Science Tools Adson Forceps, Stainless Steel, Straight, 12 cm
SDP
Catalog No. NC9602964
Encompass
Tips: Straight/ Tip Dimensions: Serrated/ Tip Width: 1.5 x 1mm/ Length: 12cm
Catalog No. NC9602964 Supplier Fine Science Tools Supplier No. 1100612
Tips: Straight/ Tip Dimensions: Serrated/ Tip Width: 1.5 x 1mm/ Length: 12cm