Learn More
American Diagnostic Corporation Adtemp 413, 30-40 Second Digital Thermometer, 25/cs
SDP
Catalog No. NC1860168
Encompass_Preferred
ADC's Adtemp 413 is a digital stick thermometer with a compact size: just under five inches long and weighing only 11 grams. Features 30 to 40 second response time, an audible tone when measurment is complete, and last reading memory.
This item is not returnable. View return policy
Catalog No. NC1860168 Supplier American Diagnostic Corporation Supplier No. 413B00
ADC's Adtemp 413 is a digital stick thermometer with a compact size: just under five inches long and weighing only 11 grams. Features 30 to 40 second response time, an audible tone when measurment is complete, and last reading memory.