Learn More
American Diagnostic Corporation Adtemp™ Thermometer Sheaths, Single Use for ADC and Most Digital Stick Thermometers, 100/PK
SDP
Catalog No. NC1430100
Encompass
Single use digital stick thermometer sheaths to reduce the risk of cross contamination.
The Adtemp™ Thermometer Sheaths features:
Non-sterile, disposable design helps eliminate cross contamination
Cost effective and easy-to-apply
Suitable for oral or rectal use
Designed for use with ADC and most comparably sized digital thermometers
This item is not returnable. View return policy
Catalog No. NC1430100 Supplier American Diagnostic Corporation Supplier No. 416100
Single use digital stick thermometer sheaths to reduce the risk of cross contamination.
The Adtemp™ Thermometer Sheaths features:
Non-sterile, disposable design helps eliminate cross contamination
Cost effective and easy-to-apply
Suitable for oral or rectal use
Designed for use with ADC and most comparably sized digital thermometers