Learn More
American Diagnostic Corporation Adtemp™ 421Tympanic IR Thermometer
SDP
Catalog No. NC1624934
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
Probe cover free design makes the Adtemp™421 tympanic thermometer ideal for home care.Measures core temperature instantly from the ear reliably and accuratelyWide Measurement Range of 32.0F to 212.0F .4F or 0.0C to 100.0C .2C depending on scale selectionProbe cover FREE design is less costly and more convenient to useLarge (6 mm) illuminated display is easy to readCompact (5 long, 1.9 oz.) ergonomic design fits comfortably in operators hand and patients earAudible fever alarm at 99.5F (37.5C)12-reading memoryComplete with lithium battery, storage/travel case, and operating instructionsMeets ASTM E1965-98 standards for accuracy
This item is not returnable. View return policy
Catalog No. NC1624934 Supplier American Diagnostic Corporation Supplier No. 421
This item has a minimum qty of 3 per supplier requirements.
Probe cover free design makes the Adtemp™421 tympanic thermometer ideal for home care.Measures core temperature instantly from the ear reliably and accuratelyWide Measurement Range of 32.0F to 212.0F .4F or 0.0C to 100.0C .2C depending on scale selectionProbe cover FREE design is less costly and more convenient to useLarge (6 mm) illuminated display is easy to readCompact (5 long, 1.9 oz.) ergonomic design fits comfortably in operators hand and patients earAudible fever alarm at 99.5F (37.5C)12-reading memoryComplete with lithium battery, storage/travel case, and operating instructionsMeets ASTM E1965-98 standards for accuracy