Learn More
Cayman Chemical ADTL-EI1712, Purity: ≥98%, CAS: 2414916-45-1, 10 mg
SDP
Catalog No. 502990493
Encompass_Preferred
A dual ERK1 and ERK5 inhibitor (IC50s = 40.43 and 64.5 nM, respectively); reduces ERK1 and ERK5 activity by 93.5% and 89.4%, respectively, but also inhibits ERK2 activity by 92.7%, in a panel of 100 kinases at 1 µM; inhibits proliferation of HL-60 and MKN74 cancer cells (IC50s = 1.26 and 2.55 µM, respectively); reduces tumor growth and intratumor phosphorylation of ERK1/2 and ERK5 in an MKN74 mouse xenograft model at 50 mg/kg per day
Catalog No. 50-299-0493 Supplier Cayman Chemical Supplier No. 3060210 MG
A dual ERK1 and ERK5 inhibitor (IC50s = 40.43 and 64.5 nM, respectively); reduces ERK1 and ERK5 activity by 93.5% and 89.4%, respectively, but also inhibits ERK2 activity by 92.7%, in a panel of 100 kinases at 1 µM; inhibits proliferation of HL-60 and MKN74 cancer cells (IC50s = 1.26 and 2.55 µM, respectively); reduces tumor growth and intratumor phosphorylation of ERK1/2 and ERK5 in an MKN74 mouse xenograft model at 50 mg/kg per day