Learn More
Medchemexpress LLC ADTL-SA1215 | 782387-91-1 | 98.96% | 657.32 | 50 MG
SDP
Catalog No. 5000350945
Encompass_Preferred
ADTL-SA1215 is a novel, first-in-class, and specific small-molecule activator of Sirtuin-3 (SIRT3). This compound is recognized for its role in modulating autophagy, particularly in the context of triple-negative breast cancer. It has demonstrated antiproliferative activity against various human cancer cell lines, including HL-60, MCF7, and MDA-MB-231. ADTL-SA1215 is intended solely for research purposes.
  • First-in-class specific small-molecule activator of SIRT3.
  • Modulates autophagy.
  • Exhibits activity in triple negative breast cancer models.
  • Demonstrates antiproliferative effects on human HL-60, MCF7, MDA-MB-231, and U-937 cell lines.
Catalog No. 50-003-50945 Supplier Medchemexpress LLC Supplier No. HY13974250MG
ADTL-SA1215 is a novel, first-in-class, and specific small-molecule activator of Sirtuin-3 (SIRT3). This compound is recognized for its role in modulating autophagy, particularly in the context of triple-negative breast cancer. It has demonstrated antiproliferative activity against various human cancer cell lines, including HL-60, MCF7, and MDA-MB-231. ADTL-SA1215 is intended solely for research purposes.
  • First-in-class specific small-molecule activator of SIRT3.
  • Modulates autophagy.
  • Exhibits activity in triple negative breast cancer models.
  • Demonstrates antiproliferative effects on human HL-60, MCF7, MDA-MB-231, and U-937 cell lines.