Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ADTL-SA1215 | 782387-91-1 | 99.0% | 657.32 | 100 MG
SDP

Catalog No. 5000348863
Encompass_Preferred

ADTL-SA1215 is a first-in-class specific small-molecule activator of SIRT3 that modulates autophagy in triple negative breast cancer. It is for research use only and not sold to patients.

  • Specific small-molecule activator of sirtuin-3 (SIRT3)
  • Modulates autophagy in triple negative breast cancer
  • For research use only
  • High purity (99.0%)
  • Solid form with light brown to brown appearance
  • Soluble in DMSO
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-48863 Supplier Medchemexpress LLC Supplier No. HY139742100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ADTL-SA1215 is a first-in-class specific small-molecule activator of SIRT3 that modulates autophagy in triple negative breast cancer. It is for research use only and not sold to patients.

  • Specific small-molecule activator of sirtuin-3 (SIRT3)
  • Modulates autophagy in triple negative breast cancer
  • For research use only
  • High purity (99.0%)
  • Solid form with light brown to brown appearance
  • Soluble in DMSO
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.